3,4-Dichloro-α-chlorobenzaldoxime
Lieferant: Apollo Scientific
OR346128-1G
OR346128-500MG
APOSOR346128-1GEA
0
EUR
APOSOR346128-1G
APOSOR346128-500MG
3,4-Dichloro-α-chlorobenzaldoxime
3,4-Dichloro-alpha-chlorobenzaldoxime
Formel:
C₇H₄Cl₃NO Molecular Weight: 224.48 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11227171 CAS-Nummer: 1056189-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...