5-Bromo-3-methylisothiazole
Lieferant: Apollo Scientific
|
Achtung
|
OR346140-1G
OR346140-5G
APOSOR346140-1GEA
201
EUR
APOSOR346140-1G
APOSOR346140-5G
5-Bromo-3-methylisothiazole
5-Bromo-3-methylisothiazole
Formel:
C₄H₄BrNS Molecular Weight: 178.06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318949 CAS-Nummer: 20493-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...