3-Methylisothiazole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR346142-1G
OR346142-500MG
APOSOR346142-1GEA
1050
EUR
APOSOR346142-1G
APOSOR346142-500MG
3-Methylisothiazole-5-carboxylic acid
3-Methylisothiazole-5-carboxylic acid
Formel:
C₅H₅NO₂S Molecular Weight: 143,17 g/mol Schmelzpunkt: 201…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110341 CAS-Nummer: 66975-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...