4-Acetoxythiophene-3-carboxylic acid methyl ester
Lieferant: Apollo Scientific
OR346146-1G OR346146-5G
APOSOR346146-1GEA 0 EUR
APOSOR346146-1G APOSOR346146-5G
4-Acetoxythiophene-3-carboxylic acid methyl ester
4-Acetoxythiophene-3-carboxylic acid methyl ester
Formel: C₈H₈O₄S
Molecular Weight: 200,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11227239
CAS-Nummer: 65369-31-5