Benzothiazol-4-ylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminobenzothiazole, Benzo[d]thiazol-4-amine

OR346148-1G OR346148-500MG
APOSOR346148-1GEA 422 EUR
APOSOR346148-1G APOSOR346148-500MG
Benzothiazol-4-ylamine
Benzothiazol-4-ylamine
Formel: C₇H₆N₂S
Molecular Weight: 150,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672135
CAS-Nummer: 1123-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR