4'-Methoxy-2'-nitroacetanilid
Lieferant: Apollo Scientific
Synonyme:
4-Acetamido-3-Nitroanisole, N-(4-Methoxy-2-nitro-phenyl)-acetamide, 4-Methoxy-2-nitroacetanilide, N-(4-Methoxy-2-nitrophenyl)acetamide
OR346152-25G
OR346152-5G
APOSOR346152-25GEA
0
EUR
APOSOR346152-25G
APOSOR346152-5G
4'-Methoxy-2'-nitroacetanilid
4'-Methoxy-2'-nitroacetanilid
Formel:
C₉H₁₀N₂O₄ Molecular Weight: 210,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017018 CAS-Nummer: 119-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...