5-Chloro-4,6-dimethylpyrimidin-2-ol
Lieferant: Apollo Scientific
OR346182-1G
OR346182-500MG
OR346182-5G
APOSOR346182-1GEA
0
EUR
APOSOR346182-1G
APOSOR346182-500MG
APOSOR346182-5G
5-Chloro-4,6-dimethylpyrimidin-2-ol
5-Chloro-4,6-dimethylpyrimidin-2-ol
Formel:
C₆H₇ClN₂O Molecular Weight: 158,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12131131 CAS-Nummer: 19573-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...