3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester
Lieferant: Apollo Scientific
Synonyme: Ethyl 1-methyl-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylate
OR346184-1G OR346184-5G
APOSOR346184-1GEA 166 EUR
APOSOR346184-1G APOSOR346184-5G
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester
3-Hydroxy-1-methyl-1H-pyrazole-4-carboxylic acid ethyl ester
Formel: C₇H₁₀N₂O₃
Molecular Weight: 170.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12131137
CAS-Nummer: 103626-03-5