(5-Bromo-benzo[b]thiophen-2-yl)methanol
Lieferant: Apollo Scientific
OR346189-1G
OR346189-5G
APOSOR346189-1GEA
0
EUR
APOSOR346189-1G
APOSOR346189-5G
(5-Bromo-benzo[b]thiophen-2-yl)methanol
(5-Bromo-benzo[b]thiophen-2-yl)methanol
Formel:
C₉H₇BrOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032279 CAS-Nummer: 13771-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...