3,3-Dibromoazepan-2-one

Lieferant: Apollo Scientific

Synonyme: 3,3-Dibromo-azepan-2-one

OR346194-1G OR346194-500MG OR346194-5G
APOSOR346194-1GEA 0 EUR
APOSOR346194-1G APOSOR346194-500MG APOSOR346194-5G
3,3-Dibromoazepan-2-one
3,3-Dibromoazepan-2-one
Formel: C₆H₉Br₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00154706
CAS-Nummer: 56987-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR