2-Brom-N,N-diethylbenzamid 97%

Lieferant: Apollo Scientific

OR3462-1G OR3462-5G
APOSOR3462-1GEA 26.1 EUR
APOSOR3462-1G APOSOR3462-5G
2-Brom-N,N-diethylbenzamid 97%
2-Brom-N,N-diethylbenzamid
Formel: C₁₁H₁₄BrNO
Molecular Weight: 256,1390 g/mol
Siedepunkt: 138…140 °C/2mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00457139
CAS-Nummer: 76041-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR