2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Verapamil Impurity K
OR346216-1G
OR346216-5G
APOSOR346216-1GEA
0
EUR
APOSOR346216-1G
APOSOR346216-5G
2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile
2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile
Formel:
C₁₃H₁₇NO₂ Molecular Weight: 219,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800230 CAS-Nummer: 20850-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...