4-Morpholin-4-ylmethylbenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Morpholinomethyl)benzoic acid

OR346277-5G
APOSOR346277-5GEA 87 EUR
APOSOR346277-5G
4-Morpholin-4-ylmethylbenzoic acid
4-Morpholin-4-ylmethylbenzoic acid
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221,26 g/mol
Schmelzpunkt: 179…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01167722
CAS-Nummer: 62642-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR