N-Hydroxypropionamidine
Lieferant: Apollo Scientific
Synonyme:
Propionamidoxime
OR346280-1G
OR346280-5G
APOSOR346280-1GEA
88.6
EUR
APOSOR346280-1G
APOSOR346280-5G
N-Hydroxypropionamidine
N-Hydroxypropionamidine
Formel:
C₃H₈N₂O Molecular Weight: 88,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426272 CAS-Nummer: 29335-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...