5-Iodo-3-methylisothiazole
Lieferant: Apollo Scientific
|
Gefahr
|
OR346311-1G
OR346311-500MG
APOSOR346311-1GEA
1420
EUR
APOSOR346311-1G
APOSOR346311-500MG
5-Iodo-3-methylisothiazole
5-Iodo-3-methylisothiazole
Formel:
C₄H₄INS Molecular Weight: 225.05 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17012839 CAS-Nummer: 20067-15-6 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...