5-Brom-2-(chlormethyl)benzoxazol 95+%
Lieferant: Apollo Scientific
OR346393-1G
APOSOR346393-1GEA
249
EUR
APOSOR346393-1G
5-Brom-2-(chlormethyl)benzoxazol 95+%
5-Bromo-2-(chloromethyl)benzoxazole
Formel:
C₈H₅BrClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD10696614 CAS-Nummer: 110704-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...