4-(tert-Butyl)-2-chloropyridine 95+%

Lieferant: Apollo Scientific

OR346413-500MG
APOSOR346413-500MGEA 192 EUR
APOSOR346413-500MG
4-(tert-Butyl)-2-chloropyridine 95+%
4-(tert-Butyl)-2-chloropyridine
Formel: C₉H₁₂ClN
Molecular Weight: 169,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12131129
CAS-Nummer: 81167-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR