(4-Chloro-1-benzothiophen-2-yl)methanol 95+%

Lieferant: Apollo Scientific

OR346415-1G OR346415-5G
APOSOR346415-1GEA 243 EUR
APOSOR346415-1G APOSOR346415-5G
(4-Chloro-1-benzothiophen-2-yl)methanol 95+%
(4-Chloro-1-benzothiophen-2-yl)methanol
Formel: C₉H₇ClOS
Molecular Weight: 198,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11841059
CAS-Nummer: 216575-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR