1,3-Dimethyl-1H-pyrazol-5-ol 95+%

Lieferant: Apollo Scientific

OR346439-1G OR346439-5G
APOSOR346439-1GEA 40.6 EUR
APOSOR346439-1G APOSOR346439-5G
1,3-Dimethyl-1H-pyrazol-5-ol 95+%
1,3-Dimethyl-1H-pyrazol-5-ol
Formel: C₅H₈N₂O
Molecular Weight: 112.1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00129673
CAS-Nummer: 5203-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR