1-Benzofuran-2-carbonitrile 95+%

Lieferant: Apollo Scientific

OR346444-1G
APOSOR346444-1GEA 58 EUR
APOSOR346444-1G
1-Benzofuran-2-carbonitrile 95+%
1-Benzofuran-2-carbonitrile
Formel: C₉H₅NO
Molecular Weight: 143,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01659779
CAS-Nummer: 41717-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR