Methyl-4-amino-3-iodbenzoat 95+%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-3-iodobenzoic acid methyl ester, Methyl 4-amino-3-iodobenzoate
OR346462-25G
APOSOR346462-25GEA
85.7
EUR
APOSOR346462-25G
Methyl-4-amino-3-iodbenzoat 95+%
Methyl-4-amino-3-iodbenzoat
Formel:
C₈H₈INO₂ Molecular Weight: 277,0600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090423 CAS-Nummer: 19718-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...