2-Methylisonicotinonitrile 95+%
Lieferant: Apollo Scientific
Synonyme:
4-Cyano-2-methylpyridine
OR346509-1G
OR346509-5G
APOSOR346509-1GEA
192
EUR
APOSOR346509-1G
APOSOR346509-5G
2-Methylisonicotinonitrile 95+%
2-Methylisonicotinonitrile
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200207 CAS-Nummer: 2214-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...