2-(4-Methoxyphenyl)pyridin 95+%
Lieferant: Apollo Scientific
Synonyme:
4-(2-Pyridyl)anisole
OR346608-1G
OR346608-5G
APOSOR346608-1GEA
84.1
EUR
APOSOR346608-1G
APOSOR346608-5G
2-(4-Methoxyphenyl)pyridin 95+%
2-(4-Methoxyphenyl)pyridine
Formel:
C₁₂H₁₁NO Molecular Weight: 185,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092333 CAS-Nummer: 5957-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...