3-Brom-5-chlortoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-chlor-5-methylbenzol
OR3493-1G
OR3493-5G
OR3493-25G
APOSOR3493-1GEA
21.9
EUR
APOSOR3493-1G
APOSOR3493-5G
APOSOR3493-25G
3-Brom-5-chlortoluol 98%
3-Brom-5-chlortoluol
Formel:
C₇H₆BrCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437596 CAS-Nummer: 329944-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...