2-Amino-4-chlorbenzothiazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR350002-100G
APOSOR350002-100GEA
177
EUR
APOSOR350002-100G
2-Amino-4-chlorbenzothiazol 98%
2-Amino-4-chlorobenzothiazole
Formel:
C₇H₅ClN₂S Molecular Weight: 184,65 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005791 CAS-Nummer: 19952-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...