1-Aminocyclohexancarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Aminocyclohexan-1-carbonsäure, Homocycloleucin

OR350005-100G OR350005-25G
APOSOR350005-100GEA 235 EUR
APOSOR350005-100G APOSOR350005-25G
1-Aminocyclohexancarbonsäure 98%
1-Aminocyclohexancarbonsäure
Formel: C₇H₁₃NO₂
Molecular Weight: 143,19 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001487
CAS-Nummer: 2756-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR