6-Bromoisoquinolin-1(2H)-one

Lieferant: Apollo Scientific
Achtung

OR350043-1G OR350043-10G
APOSOR350043-1GEA 27.7 EUR
APOSOR350043-1G APOSOR350043-10G
6-Bromoisoquinolin-1(2H)-one
6-Bromoisoquinolin-1(2H)-one
Formel: C₉H₆BrNO
Molecular Weight: 224,0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374394
CAS-Nummer: 82827-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR