6-Chlor-4(1H)-pyrimidinon 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-4(1H)-pyrimidinone, 6-Chloro-4-pyrimidinol, 4-Chloro-6-hydroxypyrimidine
OR350089-1G
OR350089-25G
OR350089-5G
APOSOR350089-1GEA
63.8
EUR
APOSOR350089-1G
APOSOR350089-25G
APOSOR350089-5G
6-Chlor-4(1H)-pyrimidinon 97%
6-Chloropyrimidin-4(1H)-one
Formel:
C₄H₃ClN₂O Molecular Weight: 130.5324 g/mol Schmelzpunkt: 194…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233957 CAS-Nummer: 4765-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...