2'-Chlor-5'-nitroacetophenon

Lieferant: Apollo Scientific
Achtung

OR350102-25G
APOSOR350102-25GEA 142 EUR
APOSOR350102-25G
2'-Chlor-5'-nitroacetophenon
2'-Chlor-5'-nitroacetophenon
Formel: C₈H₆ClNO₃
Molecular Weight: 199,5911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052836
CAS-Nummer: 23082-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR