DL-Prolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-Prolin, (±)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure, (±)-Pyrrolidin-2-carbonsäure, (±)-Azolidin-2-carbonsäure, H-DL-Pro-OH
OR350110-100G
APOSOR350110-100GEA
27.7
EUR
APOSOR350110-100G
DL-Prolin 98%
DL-Prolin
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Schmelzpunkt: 208…210 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005250 CAS-Nummer: 609-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...