Diphenylphosphinoxid 97%

Lieferant: Apollo Scientific

OR350120-100G OR350120-25G OR350120-500G
APOSOR350120-100GEA 126 EUR
APOSOR350120-100G APOSOR350120-25G APOSOR350120-500G
Diphenylphosphinoxid 97%
Diphenylphosphine oxide
Formel: C₁₂H₁₁OP
Molecular Weight: 202,20 g/mol
Siedepunkt: 105 °C/0,1mmHg
Schmelzpunkt: 56…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002079
CAS-Nummer: 4559-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR