6-Hydroxy-4-methyl-2-oxo-1,2-dihydro-3-pyridincarbonitril 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-4-methyl-2,6-pyridinediol, 3-Cyano-2,6-dihydroxy-4-picoline, 2,6-Dihydroxy-4-methylnicotinonitrile, 3-Cyano-6-hydroxy-4-methyl-2-pyridone

OR350133-100G OR350133-500G
APOSOR350133-100GEA 125 EUR
APOSOR350133-100G APOSOR350133-500G
6-Hydroxy-4-methyl-2-oxo-1,2-dihydro-3-pyridincarbonitril 95%
6-Hydroxy-4-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₇H₆N₂O₂
Molecular Weight: 150.1347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043831
CAS-Nummer: 5444-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR