Ethyl-3-hydroxy-3-phenylpropionate

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-hydroxy-3-phenylpropanoate

OR350163-5G OR350163-25G
APOSOR350163-5GEA 58 EUR
APOSOR350163-5G APOSOR350163-25G
Ethyl-3-hydroxy-3-phenylpropionate
Ethyl-3-hydroxy-3-phenylpropionate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086891
CAS-Nummer: 5764-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR