4-Ethoxyphenol
Lieferant: Apollo Scientific
OR350170-100G
OR350170-500G
APOSOR350170-100GEA
59.6
EUR
APOSOR350170-100G
APOSOR350170-500G
4-Ethoxyphenol
4-Ethoxyphenol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Siedepunkt: 121 °C Schmelzpunkt: 65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002334 CAS-Nummer: 622-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...