[4-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(1H-Pyrazol-1-yl)phenyl)boronic acid
OR350198-1G
OR350198-5G
APOSOR350198-1GEA
68.3
EUR
APOSOR350198-1G
APOSOR350198-5G
[4-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%
[4-(1H-Pyrazol-1-yl)phenyl]boronic acid
Formel:
C₉H₉BN₂O₂ Molecular Weight: 188 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739055 CAS-Nummer: 891270-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...