4-Isoxazolboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 4-Isoxazoleboronic acid pinacol ester

OR350204-1G OR350204-250MG OR350204-5G
APOSOR350204-1GEA 77 EUR
APOSOR350204-1G APOSOR350204-250MG APOSOR350204-5G
4-Isoxazolboronsäurepinakolester 98%
4-Isoxazolboronsäurepinakolester
Formel: C₉H₁₄BNO₃
Molecular Weight: 195,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06657891
CAS-Nummer: 928664-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR