Isoindoline hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR350210-5G
OR350210-10G
OR350210-1G
APOSOR350210-5GEA
46.4
EUR
APOSOR350210-5G
APOSOR350210-10G
APOSOR350210-1G
Isoindoline hydrochloride
Isoindoline hydrochloride
Formel:
C₈H₁₀ClN Schmelzpunkt: 255…256 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03502377 CAS-Nummer: 32372-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...