3-Formyl-2-picolin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Methylnicotinaldehyde
OR350218-1G
OR350218-5G
APOSOR350218-1GEA
63.8
EUR
APOSOR350218-1G
APOSOR350218-5G
3-Formyl-2-picolin 97%
3-Formyl-2-picolin
Formel:
C₇H₇NO Molecular Weight: 121,14 g/mol Dichte: 1,106 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234911 CAS-Nummer: 60032-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...