Methyldiphenylamin
Lieferant: Apollo Scientific
Synonyme:
N-Methyl-N-phenylaniline
OR350227-500G
APOSOR350227-500GEA
161
EUR
APOSOR350227-500G
Methyldiphenylamin
Methyldiphenylamin
Formel:
C₁₃H₁₃N Molecular Weight: 183,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041900 CAS-Nummer: 552-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...