3-Methylzimtsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methylcinnamsäure, 3-(3-Methylphenyl)propensäure, 3-(m-Methylphenyl)propensäure

OR350254-25G OR350254-5G
APOSOR350254-25GEA 56.7 EUR
APOSOR350254-25G APOSOR350254-5G
3-Methylzimtsäure
3-Methylzimtsäure
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Schmelzpunkt: 116…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016844
CAS-Nummer: 3029-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR