3-Brom-5-methoxyphenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-5-methoxyphenyl)boronic acid
OR3503-5G
APOSOR3503-5GEA
115
EUR
APOSOR3503-5G
3-Brom-5-methoxyphenylboronsäure 96%
3-Brom-5-methoxyphenylboronsäure
Formel:
C₇H₈BBrO₃ Molecular Weight: 230,8516 g/mol Schmelzpunkt: 224…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07369759 CAS-Nummer: 849062-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...