2-(Pyridin-4-yl)acetic acid hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-Pyridylacetic acid hydrochloride

OR350324-100G OR350324-25G
APOSOR350324-100GEA 277 EUR
APOSOR350324-100G APOSOR350324-25G
2-(Pyridin-4-yl)acetic acid hydrochloride
2-(Pyridin-4-yl)acetic acid hydrochloride
Formel: C₇H₈ClNO₂
Molecular Weight: 173.60 g/mol
Schmelzpunkt: 141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012827
CAS-Nummer: 6622-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR