2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-mercaptopyrimidin
OR350373-100G
OR350373-25G
OR350373-250G
APOSOR350373-100GEA
43.5
EUR
APOSOR350373-100G
APOSOR350373-25G
APOSOR350373-250G
2-Thioxo-2,3-dihydro-4(1H)-pyrimidinon 99%
2-Thiouracil
Formel:
C₄H₄N₂OS Molecular Weight: 128,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006039 CAS-Nummer: 141-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...