Methyl-2-((diphenylmethylene)amino)acetate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-((diphenylmethylene)amino)acetate, N-(Diphenylmethylene)glycine methyl ester, Methyl N-(diphenylmethylene)glycinate

OR350517-100G OR350517-25G
APOSOR350517-100GEA 101 EUR
APOSOR350517-100G APOSOR350517-25G
Methyl-2-((diphenylmethylene)amino)acetate
Methyl-2-((diphenylmethylene)amino)acetate
Formel: C₁₆H₁₅NO₂
Molecular Weight: 253.30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272233
CAS-Nummer: 81167-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR