4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)ethyl)piperidine
OR3511-1G
OR3511-250MG
APOSOR3511-1GEA
120
EUR
APOSOR3511-1G
APOSOR3511-250MG
4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester 95%
4-(2-(Piperidin-1-yl)ethoxy)phenylboronsäurepinakolester
Formel:
C₁₉H₃₀BNO₃ Molecular Weight: 331,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258652 CAS-Nummer: 934586-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...