3,5-Dimethyl-4-isoxazolboronsäurepinakolester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
OR3516-5G
OR3516-25G
OR3516-100G
APOSOR3516-5GEA
21.9
EUR
APOSOR3516-5G
APOSOR3516-25G
APOSOR3516-100G
3,5-Dimethyl-4-isoxazolboronsäurepinakolester 97%
3,5-Dimethyl-4-isoxazolboronsäurepinakolester
Formel:
C₁₁H₁₈BNO₃ Molecular Weight: 223.08 g/mol Schmelzpunkt: 94…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863910 CAS-Nummer: 832114-00-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...