Methyl-6-brompicolinat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-bromopicolinate, Methyl 6-bromopyridine-2-carboxylate, 6-Bromopyridine-2-carboxylic acid methyl ester, 6-Bromopicolinic acid methyl ester

OR3536-1G OR3536-5G
APOSOR3536-1GEA 21.9 EUR
APOSOR3536-1G APOSOR3536-5G
Methyl-6-brompicolinat 98%
Methyl-6-brompicolinat
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 92…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06203934
CAS-Nummer: 26218-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR