Methyl-(3-bromphenyl)acetat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromophenylacetic acid methyll ester, Methyl 3-Bromophenylacetate, Methyl 2-(3-bromophenyl)acetate

OR3537-100G OR3537-25G OR3537-5G
APOSOR3537-100GEA 248 EUR
APOSOR3537-100G APOSOR3537-25G APOSOR3537-5G
Methyl-(3-bromphenyl)acetat 98%
Methyl-(3-bromphenyl)acetat
Formel: C₉H₉BrO₂
Molecular Weight: 229.0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06858764
CAS-Nummer: 150529-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR