(4-Chlor-2-pyridinyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Chloropyridin-2-yl)methanol, 4-Chloro-2-(hydroxymethyl)pyridine

OR3538-1G OR3538-5G
APOSOR3538-1GEA 21.9 EUR
APOSOR3538-1G APOSOR3538-5G
(4-Chlor-2-pyridinyl)methanol 97%
(4-Chlor-2-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143,5709 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07437885
CAS-Nummer: 63071-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR