Methyl-2-iodnicotinat 98%

Lieferant: Apollo Scientific
Achtung

OR3543-500MG OR3543-250MG
APOSOR3543-500MGEA 250 EUR
APOSOR3543-500MG APOSOR3543-250MG
Methyl-2-iodnicotinat 98%
Methyl-2-iodnicotinat
Formel: C₇H₆INO₂
Molecular Weight: 263,03 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08689498
CAS-Nummer: 913836-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR